Learn More
Ubpbio A FRET-based fluorogenic substrate of 3L-like protease (3CLPro) from SARS viruses, including SARS-CoV-2, 5MG, CAS# 730985-86-1
Catalog No. 502339254
Encompass_Preferred
Dabcyl-KTSAVLQSGFRKME-Edans is an internally quenched fluorogenic substrate for the SARS-CoV family 3C-like protease (3CLP, also called main protease or MPro) with a Km value of 17 uM. Upon 3CLP-mediated cleavage between glutamine and serine, the resulting SGFRKME-EDANS fluorescence can be monitored at the excitation/emission wavelengths of 355/538 nm, respectively.
Catalog No. 50-233-9254 Supplier Ubpbio Supplier No. V10105
Dabcyl-KTSAVLQSGFRKME-Edans is an internally quenched fluorogenic substrate for the SARS-CoV family 3C-like protease (3CLP, also called main protease or MPro) with a Km value of 17 uM. Upon 3CLP-mediated cleavage between glutamine and serine, the resulting SGFRKME-EDANS fluorescence can be monitored at the excitation/emission wavelengths of 355/538 nm, respectively.