Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dabcyl-KTSAVLQSGFRKME-Edans TFA | 99.8% | 2081.43 | 5 MG
SDP

Catalog No. 5000423051
Encompass_Preferred

Dabcyl-KTSAVLQSGFRKME-Edans TFA is a fluorogenic peptide employed as a substrate to assess the enzymatic activities of various protease forms. This peptide also presents potential utility in the study of 2019-nCoV (COVID-19) infection.

  • Fluorogenic peptide
  • Used as a substrate to measure enzymatic activities of protease forms
  • Potential for studying 2019-nCoV (COVID-19) infection
  • Molecular weight: 2081.43 (free base)
  • Purity: 99.75%
  • Soluble in DMSO (100 mg/mL) and H2O (6.67 mg/mL) with sonication
  • Shipped at room temperature
  • Store powder at -80°C for up to 2 years, or -20°C for up to 1 year
  • Store in solvent at -80°C for up to 6 months, or -20°C for up to 1 month

Catalog No. 50-004-23051 Supplier Medchemexpress LLC Supplier No. HYP22955MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dabcyl-KTSAVLQSGFRKME-Edans TFA is a fluorogenic peptide employed as a substrate to assess the enzymatic activities of various protease forms. This peptide also presents potential utility in the study of 2019-nCoV (COVID-19) infection.

  • Fluorogenic peptide
  • Used as a substrate to measure enzymatic activities of protease forms
  • Potential for studying 2019-nCoV (COVID-19) infection
  • Molecular weight: 2081.43 (free base)
  • Purity: 99.75%
  • Soluble in DMSO (100 mg/mL) and H2O (6.67 mg/mL) with sonication
  • Shipped at room temperature
  • Store powder at -80°C for up to 2 years, or -20°C for up to 1 year
  • Store in solvent at -80°C for up to 6 months, or -20°C for up to 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.