Learn More
AdipoGen DABCYL-N-succinimidyl ester
SDP
Catalog No. 502561171
Encompass_Preferred
Chemical. CAS 146998-31-4. Formula C19H18N4O4. MW 366.37. Synthetic. N-terminal modification of peptides in automated synthesis.
Catalog No. 50-256-1171 Supplier AdipoGen Supplier No. CDXD0236M025
Chemical. CAS 146998-31-4. Formula C19H18N4O4. MW 366.37. Synthetic. N-terminal modification of peptides in automated synthesis.