Learn More
CPC Scientific DABCYL-Tyr-Val-Ala-Asp-Ala-Pro-Val-EDANS 0.5MG
SDP
Supplier:  CPC Scientific CASP023A
Encompass
Fluorogenic caspase-1 (ICE) substrate that allows a continuous assay of caspase-1 useful in the screening of inhibitory compounds (Km = 11.4 M, kcat = 0.79 s-1)SEQUENCE: DABCYL-Tyr-Val-Ala-Asp-Ala-Pro-Val-EDANSONE-LETTER SEQUENCE: DABCYL-YVADAPV-EDANSMOLECULAR FORMULA: C61H76N12O14S1MOLECULAR WEIGHT: 1233.4STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [161877-70-9]SYNONYMS: RESEARCH AREA: ApoptosisREFERENCES:M. W. Pennington and N.A. Thornberry, Peptide Res., 7, 72 (1994)M. Los et al., Nature, 375, 81 (1995)SKU(s): CASP-023A, CASP-023B
Catalog No. 50-210-8962