Learn More
Cayman Chemical Dabcyl-K-SARS-CoV-2 Replicase Polyprotein 1ab (3235-3246)-E-EDANS (trifluoroacetate salt), Purity: ≥90%, 5 mg
SDP
Catalog No. 502917528
Encompass_Preferred
A fluorogenic substrate for SARS-CoV and SARS-CoV-2 Mpro; upon enzymatic cleavage by SARS-CoV or SARS-CoV-2 Mpro at the Gln-Ser site, EDANS is separated from the Dabcyl quencher and its fluorescence can be used to quantify SARS-CoV or SARS-CoV-2 Mpro activity
Catalog No. 50-291-7528 Supplier Cayman Chemical Supplier No. 374095 MG
A fluorogenic substrate for SARS-CoV and SARS-CoV-2 Mpro; upon enzymatic cleavage by SARS-CoV or SARS-CoV-2 Mpro at the Gln-Ser site, EDANS is separated from the Dabcyl quencher and its fluorescence can be used to quantify SARS-CoV or SARS-CoV-2 Mpro activity