Learn More
Selleck Chemical LLC Dabigatran (BIBR-1048) etexilate S2154-5mg
SDP
Catalog No. 503110992
Encompass_Preferred
Dabigatran Etexilate (BIBR-1048) is the prodrug of dabigatran a potent nonpeptidic small molecule that specifically and reversibly inhibits both free and clot-bound thrombin by binding to the active site of the thrombin molecule
Catalog No. 50-311-0992 Supplier Selleck Chemical LLC Supplier No. S21545MG
Add to Cart
Add to Cart
Dabigatran Etexilate (BIBR-1048) is the prodrug of dabigatran a potent nonpeptidic small molecule that specifically and reversibly inhibits both free and clot-bound thrombin by binding to the active site of the thrombin molecule