Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dabigatran-d3 | 1246817-44-6 | 95.1% | 474.53 g/mol | C25H22D3N7O3 | 1 MG
SDP

Supplier:  Medchemexpress LLC HY10163S11MG

Encompass_Preferred

Dabigatran-d3 is the deuterium-labeled isotopologue of dabigatran provided as a solid analytical reference standard for quantitative bioanalytical assays. It is intended for use as a tracer or internal standard to improve accuracy and precision in LC-MS, GC-MS, and NMR methods, with documented storage and stability for both powder and solution forms.

  • Isotopically labeled deuterium (D3) analog for accurate quantitation.
  • Suitable as tracer and internal standard for LC-MS, GC-MS, and NMR.
  • Molecular weight 474.53 g/mol; formula C25H22D3N7O3.
  • CAS number 1246817-44-6 for unambiguous identification.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
  • Appearance: off-white to light yellow solid; packaged as 1 MG vial.

Catalog No. 50-002-49077


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.