Learn More
Medchemexpress LLC Dabigatran-d3 | 1246817-44-6 | MFCD21363573 | >99.0% | 474.53 g/mol | C25H22D3N7O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY10163S15MG
Encompass_Preferred
Dabigatran-d3 is the deuterium-labeled analogue of dabigatran designed for use as an internal standard or tracer in quantitative analytical methods. It is supplied as a solid and is intended for precise measurement of dabigatran in research and analytical workflows.
  • Deuterium-labeled internal standard for quantitative assays.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High isotopic purity for improved accuracy.
  • Solid, stable appearance for convenient handling and storage.
  • Available in multiple small-package sizes for analytical use.
Catalog No. 50-002-47623