Learn More
Medchemexpress LLC Dabigatran-d4 hydrochloride | 00-00-0 | ≥98.0% | 512.00 | C25H22D4ClN7O3 | 10 MG
SDP
Catalog No. 5000249106
Encompass_Preferred
Dabigatran-d4 hydrochloride is a deuterium-labeled analogue of dabigatran provided as the hydrochloride salt. It is intended for use as a tracer and internal standard in quantitative analytical workflows; the parent scaffold is a reversible, selective direct thrombin inhibitor with a reported Ki of 4.5 nM.
  • Deuterium-labeled for accurate mass spectrometry quantitation.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS analyses.
  • High purity (≥98.0%) for reliable analytical performance.
  • Hydrochloride salt form improves handling and stability.
  • Supports tracer studies and quantitative method development.
Catalog No. 50-002-49106 Supplier Medchemexpress LLC Supplier No. HY10163AS10MG
Dabigatran-d4 hydrochloride is a deuterium-labeled analogue of dabigatran provided as the hydrochloride salt. It is intended for use as a tracer and internal standard in quantitative analytical workflows; the parent scaffold is a reversible, selective direct thrombin inhibitor with a reported Ki of 4.5 nM.
  • Deuterium-labeled for accurate mass spectrometry quantitation.
  • Suitable as internal standard for NMR, GC-MS, and LC-MS analyses.
  • High purity (≥98.0%) for reliable analytical performance.
  • Hydrochloride salt form improves handling and stability.
  • Supports tracer studies and quantitative method development.