Learn More
Medchemexpress LLC Dabigatran etexilate | 211915-06-9 | MFCD22422841 | 99.8% | 627.73 | C34H41N7O5 | 10 MG
SDP
Catalog No. 5000246440
Encompass_Preferred
Dabigatran etexilate is the oral prodrug of the direct thrombin inhibitor dabigatran. Supplied as a solid analytical/research standard, it is intended for laboratory and analytical applications, including assay development and studies of anticoagulant activity.
  • Prodrug of dabigatran (direct thrombin inhibitor).
  • Intended for analytical and research use.
  • High purity suitable for analytical applications.
  • White to off-white powder; solid form.
  • Recommended storage: powder at -20°C or 4°C; follow product documentation.
Catalog No. 50-002-46440 Supplier Medchemexpress LLC Supplier No. HY1027410MG
Dabigatran etexilate is the oral prodrug of the direct thrombin inhibitor dabigatran. Supplied as a solid analytical/research standard, it is intended for laboratory and analytical applications, including assay development and studies of anticoagulant activity.
  • Prodrug of dabigatran (direct thrombin inhibitor).
  • Intended for analytical and research use.
  • High purity suitable for analytical applications.
  • White to off-white powder; solid form.
  • Recommended storage: powder at -20°C or 4°C; follow product documentation.