Learn More
Medchemexpress LLC Dabigatran etexilate | 211915-06-9 | MFCD22422841 | >98.0% | 627.73 g·mol⁻¹ | C34H41N7O5 | 5 MG
SDP
Catalog No. 501967997
Encompass_Preferred
Dabigatran etexilate is an orally active prodrug of dabigatran, a direct thrombin inhibitor used in anticoagulation research. Supplied as a solid reagent for preparing stock solutions, it is intended for in vitro and preclinical studies.
  • Oral prodrug of dabigatran
  • Direct thrombin inhibitor activity
  • Available in small research pack sizes (e.g., 1 mg-10 mg)
  • Molecular formula C34H41N7O5; molecular weight 627.73 g·mol⁻¹
  • Typical purity ≥98% (HPLC)
Catalog No. 50-196-7997 Supplier Medchemexpress LLC Supplier No. HY102745MG
Dabigatran etexilate is an orally active prodrug of dabigatran, a direct thrombin inhibitor used in anticoagulation research. Supplied as a solid reagent for preparing stock solutions, it is intended for in vitro and preclinical studies.
  • Oral prodrug of dabigatran
  • Direct thrombin inhibitor activity
  • Available in small research pack sizes (e.g., 1 mg-10 mg)
  • Molecular formula C34H41N7O5; molecular weight 627.73 g·mol⁻¹
  • Typical purity ≥98% (HPLC)