Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dabigatran etexilate (mesylate) | 872728-81-9 | MFCD25424070 | 99.8% | 723.8 g·mol⁻¹ | C35H45N7O8S | 200 MG
SDP

Catalog No. 5000245482
Encompass_Preferred

Dabigatran etexilate mesylate is the mesylate salt of dabigatran etexilate, an orally active prodrug of the direct thrombin inhibitor dabigatran. It is supplied as a solid for research use and is characterized by high purity and analytical documentation.

  • Oral prodrug of dabigatran, a direct thrombin inhibitor.
  • Used in research for anticoagulant and antithrombotic studies.
  • Provided as the mesylate salt in solid form suitable for analytical and formulation work.
  • High reported purity (≈99.8%) with accompanying certificate of analysis.
  • Molecular weight 723.8 g·mol⁻¹ and chemical formula C35H45N7O8S.
  • Identified by CAS number 872728-81-9 for unambiguous substance reference.

Catalog No. 50-002-45482 Supplier Medchemexpress LLC Supplier No. HY10274A200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dabigatran etexilate mesylate is the mesylate salt of dabigatran etexilate, an orally active prodrug of the direct thrombin inhibitor dabigatran. It is supplied as a solid for research use and is characterized by high purity and analytical documentation.

  • Oral prodrug of dabigatran, a direct thrombin inhibitor.
  • Used in research for anticoagulant and antithrombotic studies.
  • Provided as the mesylate salt in solid form suitable for analytical and formulation work.
  • High reported purity (≈99.8%) with accompanying certificate of analysis.
  • Molecular weight 723.8 g·mol⁻¹ and chemical formula C35H45N7O8S.
  • Identified by CAS number 872728-81-9 for unambiguous substance reference.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.