Learn More
Medchemexpress LLC Dabigatran (ethyl ester) | 429658-95-7 | MFCD16294325 | 99.0% | 499.56 g/mol | C27H29N7O3 | 10 MG
SDP
Catalog No. 5000297953
Encompass_Preferred
Dabigatran (ethyl ester) is a research-grade prodrug of dabigatran that functions as a direct thrombin inhibitor. Supplied as a powder for biochemical and pharmacology studies, it is intended for in vitro and in vivo research into anticoagulant mechanisms.
  • Direct inhibitor of thrombin activity
  • Supplied as a powder suitable for solution preparation
  • High purity (99.0%) for research applications
  • Soluble in DMSO (≈75 mg/mL) for in vitro use
  • Stable when stored under recommended conditions
  • Available in small research pack sizes for early-stage studies
Catalog No. 50-002-97953 Supplier Medchemexpress LLC Supplier No. HY1737810MG
Dabigatran (ethyl ester) is a research-grade prodrug of dabigatran that functions as a direct thrombin inhibitor. Supplied as a powder for biochemical and pharmacology studies, it is intended for in vitro and in vivo research into anticoagulant mechanisms.
  • Direct inhibitor of thrombin activity
  • Supplied as a powder suitable for solution preparation
  • High purity (99.0%) for research applications
  • Soluble in DMSO (≈75 mg/mL) for in vitro use
  • Stable when stored under recommended conditions
  • Available in small research pack sizes for early-stage studies