Learn More
TARGETMOL CHEMICALS INC Dabigatran ethyl ester hydroch
SDP
Catalog No. 502879493
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dabigatran ethyl ester hydrochloride is an effective inhibitor of ribosyl dihydronicotinic acid amide dehydrogenase (NQO2) with an IC50 value of 0.8 uM and is a thrombin inhibitor. Purity 98.7%
Catalog No. 50-287-9493 Supplier TARGETMOL CHEMICALS INC Supplier No. T1095125MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dabigatran ethyl ester hydrochloride is an effective inhibitor of ribosyl dihydronicotinic acid amide dehydrogenase (NQO2) with an IC50 value of 0.8 uM and is a thrombin inhibitor. Purity 98.7%