Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dabigatran ethyl ester hydrochloride | 211914-50-0 | MFCD09833628 | 536.03 g/mol | C27H30ClN7O3 | 10 MG
SDP

Catalog No. 5000299271
Encompass_Preferred

Dabigatran ethyl ester hydrochloride is a research-grade small-molecule inhibitor described as a thrombin inhibitor and an inhibitor of ribosyldihydronicotinamide dehydrogenase (NQO2). Supplied as a white to off-white solid in small research quantities, it is used for enzyme inhibition assays and as an impurity or derivative reference standard.

  • White to off-white solid form.
  • Molecular weight 536.03 g/mol.
  • Chemical formula C27H30ClN7O3.
  • CAS number 211914-50-0.
  • NQO2 inhibitor: IC50 = 0.8 μM.
  • Available as 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg supplies; solutions: 10 mM in DMSO.

Catalog No. 50-002-99271 Supplier Medchemexpress LLC Supplier No. HY7752110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dabigatran ethyl ester hydrochloride is a research-grade small-molecule inhibitor described as a thrombin inhibitor and an inhibitor of ribosyldihydronicotinamide dehydrogenase (NQO2). Supplied as a white to off-white solid in small research quantities, it is used for enzyme inhibition assays and as an impurity or derivative reference standard.

  • White to off-white solid form.
  • Molecular weight 536.03 g/mol.
  • Chemical formula C27H30ClN7O3.
  • CAS number 211914-50-0.
  • NQO2 inhibitor: IC50 = 0.8 μM.
  • Available as 5 mg, 10 mg, 25 mg, 50 mg, and 100 mg supplies; solutions: 10 mM in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.