Learn More
Cayman Chemical Dabigatran-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2967480-55-1, 1 mg
SDP
Catalog No. 502911394
Encompass_Preferred
An internal standard for the quantification of dabigatran by GC- or LC-MS
Catalog No. 50-291-1394 Supplier Cayman Chemical Supplier No. 357051 MG
An internal standard for the quantification of dabigatran by GC- or LC-MS