Learn More
Cayman Chemical DabIgatrn-d3 500ug
SDP
Catalog No. 502910786
Encompass_Preferred
An internal standard for the quantification of dabigatran by GC- or LC-MS
Catalog No. 50-291-0786 Supplier Cayman Chemical Supplier No. 35704500 UG
An internal standard for the quantification of dabigatran by GC- or LC-MS