Learn More
Cayman Chemical DabIgatrn etexIlate 10mg
SDP
Catalog No. 502966637
Encompass_Preferred
A prodrug form of dabigatran; inhibits thrombin-induced platelet aggregation in isolated rabbit platelets (IC50 = 0.337 µM); inhibits thrombus formation in a rat model of venous thrombosis (ED50 = 7.8 µmol/kg)
Catalog No. 50-296-6637 Supplier Cayman Chemical Supplier No. 1713110 MG
A prodrug form of dabigatran; inhibits thrombin-induced platelet aggregation in isolated rabbit platelets (IC50 = 0.337 µM); inhibits thrombus formation in a rat model of venous thrombosis (ED50 = 7.8 µmol/kg)