Learn More
Cayman Chemical Dabigatran etexilate, Purity: ≥95%, CAS: 211915-06-9, 50 mg
SDP
Catalog No. 502966605
Encompass_Preferred
A prodrug form of dabigatran; inhibits thrombin-induced platelet aggregation in isolated rabbit platelets (IC50 = 0.337 µM); inhibits thrombus formation in a rat model of venous thrombosis (ED50 = 7.8 µmol/kg)
Catalog No. 50-296-6605 Supplier Cayman Chemical Supplier No. 1713150 MG
A prodrug form of dabigatran; inhibits thrombin-induced platelet aggregation in isolated rabbit platelets (IC50 = 0.337 µM); inhibits thrombus formation in a rat model of venous thrombosis (ED50 = 7.8 µmol/kg)