Learn More
Cayman Chemical DabIgatrn etexIlate-d13 1mg
SDP
Catalog No. 502967405
Encompass_Preferred
An internal standard for the quantification of dabigatran etexilate by GC- or LC-MS
Catalog No. 50-296-7405 Supplier Cayman Chemical Supplier No. 184511 MG
Add to Cart
Add to Cart
An internal standard for the quantification of dabigatran etexilate by GC- or LC-MS