Learn More
Cayman Chemical DabIgatrn etexIlate-d13 500ug
SDP
Catalog No. 502964960
Encompass_Preferred
An internal standard for the quantification of dabigatran etexilate by GC- or LC-MS
Catalog No. 50-296-4960 Supplier Cayman Chemical Supplier No. 18451500 UG
Add to Cart
Add to Cart
An internal standard for the quantification of dabigatran etexilate by GC- or LC-MS