Learn More
Medchemexpress LLC HY-14660 10mg Medchemexpress, Dabrafenib CAS:1195765-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1466010MG
Encompass_Preferred
Medchemexpress, HY-14660 10mg Dabrafenib CAS:1195765-45-7 Dabrafenib (GSK2118436A) is an ATP-competitive inhibitor of Raf with IC50s of 5 nM and 0.6 nM for C-Raf and B-RafV600E, respectively[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9058