Learn More
Medchemexpress LLC HY-14660 100mg , Dabrafenib GSK2118436A;GSK2118436 CAS:1195765-45-7 Purity:98%
SDP
Catalog No. 501492693
Encompass_Preferred
Medchemexpress, HY-14660 100mg Dabrafenib GSK2118436A;GSK2118436 CAS:1195765-45-7 Formula:C23H20F3N5O2S2 IC50: 0.6 nM (BRAF V600E ), 5 nM (CRAF) Purity:98% Dabrafenib is an ATP-competitive inhibitor of BRAF with IC 50 s of 5 nM and 0.6 nM for CRAF and BRAF V600E , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2693 Supplier Medchemexpress LLC Supplier No. HY14660100MG
Medchemexpress, HY-14660 100mg Dabrafenib GSK2118436A;GSK2118436 CAS:1195765-45-7 Formula:C23H20F3N5O2S2 IC50: 0.6 nM (BRAF V600E ), 5 nM (CRAF) Purity:98% Dabrafenib is an ATP-competitive inhibitor of BRAF with IC 50 s of 5 nM and 0.6 nM for CRAF and BRAF V600E , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.