Learn More
Medchemexpress LLC HY-14660 200mg , Dabrafenib CAS:1195765-45-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14660200MG
Encompass_Preferred
HY-14660 200mg Dabrafenib CAS: 1195765-45-7 Dabrafenib (GSK2118436A) is an ATP-competitive inhibitor of Raf with IC50s of 5 nM and 0.6 nM for C-Raf and B-RafV600E, respectively[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9002