Learn More
Enzo Life Sciences Dabrafenib (free base) (25mg). CAS: 1195765-45-7
SDP
Catalog No. 502013570
Encompass_Preferred
Dabrafenib is a mutant BRAFV600 specific inhibitor with IC50 of 0.8 nM in cell-free assays, with 4- and 6-fold less potency against B-Raf(wt) and c-Raf, respectively. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3570 Supplier Enzo Life Sciences Supplier No. ENZCHM2190025
Dabrafenib is a mutant BRAFV600 specific inhibitor with IC50 of 0.8 nM in cell-free assays, with 4- and 6-fold less potency against B-Raf(wt) and c-Raf, respectively. Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.