Learn More
Selleck Chemical LLC Dabrafenib (GSK2118436) 10mM/1mL 1195765-45-7
SDP
Supplier:  Selleck Chemical LLC S280710MM/1ML
Encompass_Preferred
Dabrafenib (GSK2118436) is a mutant BRAFV600 specific inhibitor with IC50 of 0.7 nM in cell-free assays, with 7- and 9-fold less potency against B-Raf(wt) and c-Raf, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0621920