Learn More
TARGETMOL CHEMICALS INC DABRAFENIB MESYLATE 100MG
SDP
Catalog No. 502576241
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dabrafenib Mesylate (GSK2118436 Mesylate) is a B-Raf inhibitor(IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf respectively). purity: 99%
Catalog No. 50-257-6241 Supplier TARGETMOL CHEMICALS INC Supplier No. T8474100MG
Also available in 5 mg 10 mg 25 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dabrafenib Mesylate (GSK2118436 Mesylate) is a B-Raf inhibitor(IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf respectively). purity: 99%