Learn More
Apexbio Technology LLC Dabrafenib Mesylate (GSK-2118436), 100mg
SDP
Supplier:  Apexbio Technology LLC A3347100
Encompass_Preferred
GSK2118436 is a selective BRAF V600E inhibitor. BRAF encodes a proto-oncogene B-Raf also known as serine/threonine protein kinase B-Raf. It is critical in regulating the MAPK/ERK signaling pathway. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4798