Learn More
Apexbio Technology LLC Dabrafenib Mesylate (GSK-2118436) 1195768-06-9 10mg
SDP
Catalog No. 5000063516
Encompass_Preferred
Dabrafenib Mesylate (GSK-2118436 CAS 1195768-06-9) is a potent orally active inhibitor that specifically targets the mutant BRAF kinase variant V600E BRAF functions as a serine/threonine kinase essential in the MAPK/ERK signaling cascade frequently mutated in various human cancers Dabrafenib selectively inhibits BRAF V600E (IC50 0 8 nM) with lower activity against wild-type B-Raf and c-Raf By binding and disrupting BRAF V600E activity dabrafenib attenuates downstream MAPK/ERK activation leading to reduced cellular proliferation and tumorigenicity This agent is useful for investigating oncogenic pathways and was FDA-approved for metastatic melanoma with BRAF V600E mutation
Catalog No. 50-000-63516 Supplier Apexbio Technology LLC Supplier No. A334710
Add to Cart
Add to Cart
Dabrafenib Mesylate (GSK-2118436 CAS 1195768-06-9) is a potent orally active inhibitor that specifically targets the mutant BRAF kinase variant V600E BRAF functions as a serine/threonine kinase essential in the MAPK/ERK signaling cascade frequently mutated in various human cancers Dabrafenib selectively inhibits BRAF V600E (IC50 0 8 nM) with lower activity against wild-type B-Raf and c-Raf By binding and disrupting BRAF V600E activity dabrafenib attenuates downstream MAPK/ERK activation leading to reduced cellular proliferation and tumorigenicity This agent is useful for investigating oncogenic pathways and was FDA-approved for metastatic melanoma with BRAF V600E mutation