Learn More
Medchemexpress LLC HY-14660A 10mg Medchemexpress, Dabrafenib (Mesylate) CAS:1195768-06-9 Purity:>98%
SDP
Catalog No. 502029059
Encompass_Preferred
Medchemexpress, HY-14660A 10mg Dabrafenib (Mesylate) CAS:1195768-06-9 Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9059 Supplier Medchemexpress LLC Supplier No. HY14660A10MG
Medchemexpress, HY-14660A 10mg Dabrafenib (Mesylate) CAS:1195768-06-9 Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.