Learn More
Medchemexpress LLC Dabrafenib Mesylate | 1195768-06-9 | 99.9% | 615.67 | 100 MG
SDP
Catalog No. 5000360670
Encompass_Preferred
Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. It is intended for research use only.
  • Inhibits more than 90% of cell growth in NRAS mutant clones when combined with GSK1120212.
  • Reduces S6P phosphorylation in A375 cells.
  • Suppresses polyp-mediated vascular barrier permeability, inflammatory biomarker upregulation, leukocyte adhesion/migration, and activation/production of NF-κB, TNF-α, and IL-6.
  • Inhibits HMGB1 release and downregulates HMGB1-dependent inflammatory responses by enhancing cell adhesion molecule (CAMs) expressions in human endothelial cells.
  • In vivo studies showed that dabrafenib-treated females had mostly immature reproductive tracts with no evidence of ovulation, but keratinized and histologically open vaginas.
Catalog No. 50-003-60670 Supplier Medchemexpress LLC Supplier No. HY14660A100MG
Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. It is intended for research use only.
  • Inhibits more than 90% of cell growth in NRAS mutant clones when combined with GSK1120212.
  • Reduces S6P phosphorylation in A375 cells.
  • Suppresses polyp-mediated vascular barrier permeability, inflammatory biomarker upregulation, leukocyte adhesion/migration, and activation/production of NF-κB, TNF-α, and IL-6.
  • Inhibits HMGB1 release and downregulates HMGB1-dependent inflammatory responses by enhancing cell adhesion molecule (CAMs) expressions in human endothelial cells.
  • In vivo studies showed that dabrafenib-treated females had mostly immature reproductive tracts with no evidence of ovulation, but keratinized and histologically open vaginas.