Learn More
Medchemexpress LLC Dabrafenib (mesylate) | 1195768-06-9 | 99.9% | 615.67 | 50 MG
SDP
Catalog No. 5000360913
Encompass_Preferred
Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. It is intended for research use.
  • Inhibits over 90% of cell growth in NRAS mutant clones
  • Reduces S6P phosphorylation in A375 cells
  • Suppresses PolyP-mediated vascular barrier permeability
  • Downregulates HMGB1-dependent inflammatory responses
  • Enhances the expression of cell adhesion molecules (CAMs) in human endothelial cells
Catalog No. 50-003-60913 Supplier Medchemexpress LLC Supplier No. HY14660A50MG
Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. It is intended for research use.
  • Inhibits over 90% of cell growth in NRAS mutant clones
  • Reduces S6P phosphorylation in A375 cells
  • Suppresses PolyP-mediated vascular barrier permeability
  • Downregulates HMGB1-dependent inflammatory responses
  • Enhances the expression of cell adhesion molecules (CAMs) in human endothelial cells