Learn More
Selleck Chemical LLC Dabrafenib Mesylate-25mg
SDP
Supplier:  Selleck Chemical LLC S506925MG
Encompass_Preferred
Dabrafenib Mesylate (GSK2118436) is the mesylate salt form of dabrafenib, an orally bioavailable inhibitor of B-raf (BRAF) protein with IC50s of 0.8 nM, 3.2 nM and 5 nM for B-Raf (V600E), B-Raf (WT) and C-Raf, respectively.
Catalog No. 50-239-0047