Learn More
Medchemexpress LLC HY-14660A 500mg , Dabrafenib (Mesylate) CAS:1195768-06-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14660A500MG
Encompass_Preferred
HY-14660A 500mg Dabrafenib (Mesylate) CAS: 1195768-06-9 Dabrafenib Mesylate is a potent and selective Raf kinase inhibitor with IC50s of 0.6 and 5.0 nM for RafV600E and c-Raf, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0878