Learn More
Selleck Chemical LLC Dabrafenib Mesylate S5069-1g
SDP
Catalog No. 503112457
Encompass_Preferred
Dabrafenib Mesylate (GSK2118436) is the mesylate salt form of dabrafenib an orally bioavailable inhibitor of B-raf (BRAF) protein with IC50s of 0 8 nM 3 2 nM and 5 nM for B-Raf (V600E) B-Raf (WT) and C-Raf respectively
Catalog No. 50-311-2457 Supplier Selleck Chemical LLC Supplier No. S50691G
Dabrafenib Mesylate (GSK2118436) is the mesylate salt form of dabrafenib an orally bioavailable inhibitor of B-raf (BRAF) protein with IC50s of 0 8 nM 3 2 nM and 5 nM for B-Raf (V600E) B-Raf (WT) and C-Raf respectively