Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dabsyl-L-proline 98.0+%, TCI America™
SDP

Catalog No. D1458100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1458100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1458100MG Supplier TCI America Supplier No. D1458100MG

Chemical Identifiers

CAS 89131-09-9
Molecular Formula C19H22N4O4S
Molecular Weight (g/mol) 402.469
MDL Number MFCD00059729
InChI Key IIHLDMYXACDQGQ-SFHVURJKSA-N
Synonym 4-Dimethylaminoazobenzene-4′C-sulfonyl-L-proline, Dbs-Pro-OH
PubChem CID 14048715
IUPAC Name (2S)-1-[4-[[4-(dimethylamino)phenyl]diazenyl]phenyl]sulfonylpyrrolidine-2-carboxylic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)N3CCCC3C(=O)O

Specifications

Melting Point 172°C
Quantity 100 mg
Formula Weight 402.47
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dabsyl-L-proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.