Learn More
Medchemexpress LLC HY-117071 5mg Medchemexpress, Dabuzalgron CAS:219311-44-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1170715MG
Encompass_Preferred
Medchemexpress, HY-117071 5mg Dabuzalgron CAS:219311-44-1 Dabuzalgron (Ro 115-1240) is an orally active and selective α-1A adrenergic receptor agonist for the treatment of urinary incontinence. Dabuzalgron protects against Doxorubicin-induced cardiotoxicity by preserving mitochondrial function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9451