Learn More
Selleck Chemical LLC Dac51-5mg
SDP
Catalog No. 502389740
Encompass_Preferred
Dac51 is a potent FTO inhibitor with an IC50 of 0.4 μM for inhibiting FTO demethylation.
Catalog No. 50-238-9740 Supplier Selleck Chemical LLC Supplier No. S98765MG
Dac51 is a potent FTO inhibitor with an IC50 of 0.4 μM for inhibiting FTO demethylation.