Learn More
Medchemexpress LLC HY-B0078 1g , Dacarbazine CAS:4342-03-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB00781G
Encompass_Preferred
HY-B0078 1g Dacarbazine CAS: 4342-03-4 Dacarbazine is a cell cycle nonspecific antineoplastic alkylating agent. Dacarbazine inhibits T and B lymphoblastic response, with IC50 values of 50 and 10 μg/ml, respectively. Dacarbazine can be used for the research of metastatic malignant melanoma. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0281