Learn More
Apexbio Technology LLC Dacarbazine 4342-03-4 200mg
SDP
Catalog No. 502240449
Encompass_Preferred
Dacarbazine (CAS 4342-03-4) is a triazene-based alkylating agent widely used in biomedical research and oncology Upon metabolic activation dacarbazine generates reactive methylating species that transfer methyl groups primarily to the O6 and N7 positions of guanine bases within DNA This alkylation promotes DNA strand cross-linking and mispairing ultimately inducing apoptosis in rapidly dividing cells Dacarbazine is commonly applied in studies modeling malignant melanoma Hodgkin lymphoma sarcoma and pancreatic islet cell carcinoma Its mechanism of DNA damage and cytotoxicity also informs research into chemoresistance and combination chemotherapy regimens
This item is not returnable. View return policy
Catalog No. 50-224-0449 Supplier Apexbio Technology LLC Supplier No. A2197200
Dacarbazine (CAS 4342-03-4) is a triazene-based alkylating agent widely used in biomedical research and oncology Upon metabolic activation dacarbazine generates reactive methylating species that transfer methyl groups primarily to the O6 and N7 positions of guanine bases within DNA This alkylation promotes DNA strand cross-linking and mispairing ultimately inducing apoptosis in rapidly dividing cells Dacarbazine is commonly applied in studies modeling malignant melanoma Hodgkin lymphoma sarcoma and pancreatic islet cell carcinoma Its mechanism of DNA damage and cytotoxicity also informs research into chemoresistance and combination chemotherapy regimens