Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dacarbazine, Fisher BioReagents

Catalog No. BP25135 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP25135 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP25135

missing translation for 'validMainframeMsg'

Dacarbazine is an alkylating agent used in cancer research. Antineoplastic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3-4-4342
Molecular Formula C6H10N6O
Molecular Weight (g/mol) 182.19
InChI Key OMJKFYKNWZZKTK-UXBLZVDNSA-N
Synonym dacarbazine, biocarbazine, deticene, dtic-dome, biocarbazine r, decarbazine, dtic, icdt, dacarbazinum inn-latin, dacarbazino inn-spanish
PubChem CID 5353562
IUPAC Name (5E)-5-(dimethylaminohydrazinylidene)imidazole-4-carboxamide
SMILES CN(C)NN=C1C(=NC=N1)C(=O)N

Specifications

Melting Point 202°C
Color White
Packaging Amber Glass
Quantity 5 g
Merck Index 15, 2795
Formula Weight 182.2
Physical Form Powder or Solid
Chemical Name or Material Dacarbazine
Safety handling Check Alert storage code Red
Recommended Storage 4° C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.