Learn More
Aviva Systems Biology Corp DACH1 Antibody - C-terminal region (ARP39788_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39788_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DACH1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DACH1. Alternate names: DACH, FLJ10138ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-829