Learn More
Aviva Systems Biology Corp DACH1 Antibody - N-terminal region (ARP39787_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39787_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DACH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DACH1. Alternate names: DACH, FLJ10138ApplicationWB. Species reactivity: Human, Mouse, Pig, Yeast
Catalog No. 50-014-57179