Learn More
Aviva Systems Biology Corp DACH2 Antibody - C-terminal region (ARP32425_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32425_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DACH2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human DACH2. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54583