Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Daclatasvir | 1009119-64-5 | MFCD18074519 | 98.7% | 738.88 g/mol | C40H50N8O6 | 200 MG
SDP

Catalog No. 5000242585
Encompass_Preferred

Daclatasvir is a potent NS5A inhibitor used as a research reagent to study hepatitis C virus replication and related pharmacology. The compound shows picomolar-range antiviral activity across multiple HCV genotypes and also inhibits OATP1B and OATP1B3 transporters. It is supplied as a high-purity solid and is commonly provided in solution formats for in vitro assays.

  • Potent NS5A inhibition with EC50s in the picomolar range.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes for small- and large-scale experiments.
  • Soluble in DMSO and commonly supplied as 10 mM solution for convenience.
  • Also inhibits OATP1B and OATP1B3, relevant for transporter studies.
  • Molecular weight 738.88 g/mol for formula-based calculations.

Catalog No. 50-002-42585 Supplier Medchemexpress LLC Supplier No. HY10466200MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Daclatasvir is a potent NS5A inhibitor used as a research reagent to study hepatitis C virus replication and related pharmacology. The compound shows picomolar-range antiviral activity across multiple HCV genotypes and also inhibits OATP1B and OATP1B3 transporters. It is supplied as a high-purity solid and is commonly provided in solution formats for in vitro assays.

  • Potent NS5A inhibition with EC50s in the picomolar range.
  • High purity suitable for biochemical and cellular assays.
  • Available in multiple pack sizes for small- and large-scale experiments.
  • Soluble in DMSO and commonly supplied as 10 mM solution for convenience.
  • Also inhibits OATP1B and OATP1B3, relevant for transporter studies.
  • Molecular weight 738.88 g/mol for formula-based calculations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.