Learn More
Selleck Chemical LLC Daclatasvir (BMS-790052) S1482-5mg
SDP
Catalog No. 503111343
Encompass_Preferred
Daclatasvir (BMS-790052 EBP883) is a highly selective inhibitor of HCV NS5A with EC50 of 9-50 pM for a broad range of HCV replicon genotypes and the JFH-1 genotype 2a infectious virus in cell culture Phase 3
Catalog No. 50-311-1343 Supplier Selleck Chemical LLC Supplier No. S14825MG
Daclatasvir (BMS-790052 EBP883) is a highly selective inhibitor of HCV NS5A with EC50 of 9-50 pM for a broad range of HCV replicon genotypes and the JFH-1 genotype 2a infectious virus in cell culture Phase 3