Learn More
Apexbio Technology LLC Daclatasvir (BMS-790052), 25mg. Cas: 1009119-64-5; 1214735-16-6 MFCD: MFCD17129086. HCV NS5A inhibitor
SDP
Supplier:  Apexbio Technology LLC A561825
Encompass_Preferred
Daclatasvir, formerly known as BMS-790052, is a potent NS5A inhibitor with EC50 values varying from 9 to 146 pM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4957
 Restricted Item