Learn More
Abcam Daclatasvir (BMS-790052), HCV NS5A protein inhibitor, 25MG
Supplier:  Abcam AB23150725MG
Encompass_Preferred
MW 738.9 Da, Purity =97%. Daclatasvir a potent, direct acting, small molecule inhbitor of HCV NS5A protein. It inhbibits replicons expressing a wide range of HCV genotypes with EC₅₀ values in the pM range (9 and 71 pM against the 1b and 2a genotypes, respectively). It also inhibits infection by the JFH-1 genotype 2a virus in cultured cells. Daclatasvir is thought to inhibit relication of the HCV viral genome by affecting the subcellular localization of HCV NS5A and preventing its incorporation into relication complexes. In HCV-infected Huh7 cells, Daclatasvir (1 nM) inhibits viral RNA synthesis and viron assembly. Daclatasvir displays moderate activity against the OAT1B1 (IC₅₀ = 1.5 μM) and OAT1B3 (IC₅₀ = 3.27 μM) organic anion transporters.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2640