Learn More
Medchemexpress LLC Daclatasvir dihydrochloride | 1009119-65-6 | MFCD25541736 | 99.7% | 811.80 g·mol⁻¹ | C40H52Cl2N8O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10465R25MG
Encompass_Preferred
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent HCV NS5A inhibitor supplied as a research-grade reference standard in a 25 mg sample. It is intended for in vitro screening, mechanistic studies of NS5A, and analytical reference work. Key identifiers include CAS 1009119-65-6, molecular formula C40H52Cl2N8O6, molecular weight 811.80 g·mol⁻1, and typical stated purity of 99.65%.
  • High stated purity suitable for analytical reference.
  • Provided as a 25 mg research sample.
  • Identified by CAS 1009119-65-6 and molecular formula C40H52Cl2N8O6.
  • Molecular weight 811.80 g·mol⁻1.
  • Intended for in vitro and analytical applications.
Catalog No. 50-002-44049