Learn More
Medchemexpress LLC Daclatasvir dihydrochloride | 1009119-65-6 | MFCD25541736 | 99.8% | 811.80 | C40H52Cl2N8O6 | 1 G
SDP
Catalog No. 5000243303
Encompass_Preferred
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent hepatitis C virus (HCV) NS5A inhibitor supplied for laboratory research. The material is provided as a solid powder with documented analytical support for use in in vitro and biochemical studies targeting HCV replication pathways.
  • High purity suitable for research applications.
  • Provided as a solid dihydrochloride salt for stability.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for traceability.
  • Available in multiple sizes and solution formats for assay flexibility.
  • Molecular weight 811.80 and CAS 1009119-65-6 included for identification.
Catalog No. 50-002-43303 Supplier Medchemexpress LLC Supplier No. HY104651G
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent hepatitis C virus (HCV) NS5A inhibitor supplied for laboratory research. The material is provided as a solid powder with documented analytical support for use in in vitro and biochemical studies targeting HCV replication pathways.
  • High purity suitable for research applications.
  • Provided as a solid dihydrochloride salt for stability.
  • Supplied with datasheet, certificate of analysis, and safety data sheet for traceability.
  • Available in multiple sizes and solution formats for assay flexibility.
  • Molecular weight 811.80 and CAS 1009119-65-6 included for identification.